siRNA Details
Gene Symbol: Slc4a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021530.2 | NP_067505.2 | 15 | 2117 | ||
GenBank mRNA | AF224508.1 | - | 15 | 2063 | ||
AK034237.1 | - | 15 | 2119 | |||
AK047863.1 | - | 8 | 1051 | |||
AK171610.1 | - | 14 | 1893 | |||
AK173014.1 | - | 15 | 1951 | |||
BC030388.1 | - | 15 | 2079 | 5401143 |
