siRNA Details
Gene Symbol: Tmem8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021793.2 | NP_068565.1 | 5 | 960 | ||
GenBank mRNA | AB045293.1 | - | 5 | 959 | ||
AK044384.1 | - | 5 | 961 | |||
AK079893.1 | - | 1 | 107 | |||
AK198386.1 | - | 1 | 116 | |||
BC005722.1 | - | 4 | 836 | 3595205 |
