siRNA Details
Gene Symbol: Trpv4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022017.3 | NP_071300.2 | 6 | 1149 | ||
GenBank mRNA | AB021875.2 | - | 5 | 1076 | ||
AF208026.1 | - | 6 | 1117 | |||
AF263522.1 | - | 5 | 1107 | |||
AF279672.1 | - | 6 | 1133 | |||
AJ296078.1 | - | 5 | 1068 | |||
AK083223.1 | - | 6 | 1120 | |||
AK158652.1 | - | 2 | 1002 | |||
BC127052.1 | - | 6 | 1114 | 40109678 |
