siRNA Details
Gene Symbol: Trpv4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022017.3 | NP_071300.2 | 8 | 1463 | ||
GenBank mRNA | AB021875.2 | - | 7 | 1390 | ||
AF208026.1 | - | 8 | 1431 | |||
AF263522.1 | - | 7 | 1421 | |||
AF279672.1 | - | 8 | 1447 | |||
AJ296078.1 | - | 7 | 1382 | |||
AK083223.1 | - | 8 | 1434 | |||
AK158652.1 | - | 4 | 1316 | |||
BC127052.1 | - | 8 | 1428 | 40109678 |
