siRNA Details
Gene Symbol: Sv2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022030.3 | NP_071313.1 | 7 | 1630 | ||
GenBank mRNA | AK028318.1 | - | 7 | 1575 | ||
AK080502.1 | - | 7 | 1583 | |||
AK080525.1 | - | 7 | 1672 | |||
AK122360.1 | - | 7 | 1666 | |||
BC046587.1 | - | 7 | 1630 | 6493509 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK158936.1 | - | 2512 | Chr. - - [-] on Build |
