siRNA Details
Gene Symbol: Tsc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289575.1 | NP_001276504.1 | 17, 18 | 2418 | ||
NM_001289576.1 | NP_001276505.1 | 17, 18 | 2400 | |||
NM_022887.4 | NP_075025.2 | 17, 18 | 2386 | |||
GenBank mRNA | AB047561.1 | - | 15, 16 | 2185 | ||
AJ271912.1 | - | 17, 18 | 2410 | |||
AK030318.1 | - | 17, 18 | 2383 | |||
AK122229.1 | - | 3, 4 | 710 | |||
AK147428.1 | - | 17, 18 | 2391 | |||
BC052399.1 | - | 16, 17 | 2339 | 5720967 |
