siRNA Details
Gene Symbol: Tsc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289575.1 | NP_001276504.1 | 18 | 2536 | ||
NM_001289576.1 | NP_001276505.1 | 18 | 2518 | |||
NM_022887.4 | NP_075025.2 | 18 | 2504 | |||
GenBank mRNA | AB047561.1 | - | 16 | 2303 | ||
AJ271912.1 | - | 18 | 2528 | |||
AK030318.1 | - | 18 | 2501 | |||
AK122229.1 | - | 4 | 828 | |||
AK147428.1 | - | 18 | 2509 | |||
BC052399.1 | - | 17 | 2457 | 5720967 |
