siRNA Details
Gene Symbol: Tmem176b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164207.1 | NP_001157679.1 | 7 | 1158 | ||
NM_001164208.1 | NP_001157680.1 | 7 | 806 | |||
NM_001164209.2 | NP_001157681.1 | 8 | 1014 | |||
NM_001286651.1 | NP_001273580.1 | 8 | 1011 | |||
NM_001286652.1 | NP_001273581.1 | 7 | 918 | |||
NM_023056.4 | NP_075543.1 | 7 | 1407 | |||
GenBank mRNA | AB031386.1 | - | 7 | 967 | ||
AK007408.1 | - | 7 | 813 | |||
AK156042.1 | - | 7 | 1159 | |||
BC019539.1 | - | 6 | 776 | 4237639 | ||
BC022166.1 | - | 7 | 1422 | 3582123 | ||
BE457679.1 | - | 1 | 261 | |||
DV059241.1 | - | 7 | 883 |
