siRNA Details
Gene Symbol: Tmem176b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164207.1 | NP_001157679.1 | 5 | 885 | ||
NM_001164208.1 | NP_001157680.1 | 5 | 533 | |||
NM_001164209.2 | NP_001157681.1 | 6 | 741 | |||
NM_001286651.1 | NP_001273580.1 | 6 | 738 | |||
NM_001286652.1 | NP_001273581.1 | 5 | 645 | |||
NM_023056.4 | NP_075543.1 | 5 | 1134 | |||
GenBank mRNA | AB031386.1 | - | 5 | 694 | ||
AF115426.1 | - | 5 | 606 | |||
AK007408.1 | - | 5 | 540 | |||
AK156042.1 | - | 5 | 886 | |||
BC019539.1 | - | 4 | 503 | 4237639 | ||
BC022166.1 | - | 5 | 1149 | 3582123 | ||
BQ571258.1 | - | 5 | 525 | |||
CT010176.1 | - | 4 | 498 | |||
DV059241.1 | - | 5 | 609 |
