siRNA Details
Gene Symbol: Tmem57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025382.6 | NP_079658.2 | 6 | 1026 | ||
GenBank mRNA | AK138874.1 | - | 1 | 674 | ||
AY364165.1 | - | 6 | 811 | |||
AY846873.1 | - | 6 | 1027 | |||
AY846874.1 | - | 6 | 1027 | |||
BC019186.1 | - | 6 | 995 | |||
BC043306.1 | - | 3 | 458 | |||
BC054355.1 | - | 7 | 939 | |||
BC054769.1 | - | 6 | 919 | 6415086 |
