siRNA Details
Gene Symbol: Nip7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164472.1 | NP_001157944.1 | 2, 3 | 193 | ||
NM_025391.2 | NP_079667.2 | 2, 3 | 193 | |||
GenBank mRNA | AK003739.1 | - | 2, 3 | 193 | ||
AK006057.1 | - | 2, 3 | 172 | |||
AK014290.1 | - | 2, 3 | 152 | |||
BC003972.1 | - | 2, 3 | 180 | 3488726 |
