siRNA Details
Gene Symbol: Aptx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025444.3 | NP_001020615.1 | 6, 7 | 815 | ||
NM_001025445.2 | NP_001020616.1 | 6, 7 | 810 | |||
NM_025545.4 | NP_079821.3 | 6, 7 | 950 | |||
GenBank mRNA | AK005286.1 | - | 5, 6 | 767 | ||
AK010516.1 | - | 6, 7 | 797 | |||
AK017522.1 | - | 5, 6 | 778 | |||
AK077351.1 | - | 6, 7 | 951 | |||
AK088928.1 | - | 5, 6 | 500 | |||
AY040780.1 | - | 6, 7 | 961 | |||
AY208844.1 | - | 6, 7 | 962 | |||
BC021872.1 | - | 6, 7 | 808 | 3994375 | ||
BC068309.1 | - | 5, 6 | 723 | |||
BC071213.1 | - | 6, 7 | 802 |
