siRNA Details
Gene Symbol: Aptx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025444.3 | NP_001020615.1 | 3, 4 | 230 | ||
NM_001025445.2 | NP_001020616.1 | 3, 4 | 225 | |||
NM_025545.4 | NP_079821.3 | 3, 4 | 365 | |||
GenBank mRNA | AK005286.1 | - | 2, 3 | 182 | ||
AK010516.1 | - | 3, 4 | 212 | |||
AK017522.1 | - | 2, 3 | 195 | |||
AK077351.1 | - | 3, 4 | 366 | |||
AY040780.1 | - | 3, 4 | 376 | |||
AY208844.1 | - | 3, 4 | 377 | |||
BB860888.1 | - | 3, 4 | 234 | |||
BC021872.1 | - | 3, 4 | 223 | 3994375 | ||
BC068309.1 | - | 2, 3 | 138 | |||
BC071213.1 | - | 3, 4 | 217 |
