siRNA Details
Gene Symbol: Aptx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025444.3 | NP_001020615.1 | 2, 3 | 201 | ||
NM_025545.4 | NP_079821.3 | 2, 3 | 336 | |||
GenBank mRNA | AK005286.1 | - | 1, 2 | 153 | ||
AK077351.1 | - | 2, 3 | 337 | |||
AK088928.1 | - | 2, 3 | 171 | |||
AY040780.1 | - | 2, 3 | 347 | |||
AY208844.1 | - | 2, 3 | 348 | |||
BB860888.1 | - | 2, 3 | 205 | |||
BC021872.1 | - | 2, 3 | 194 | 3994375 | ||
BC071213.1 | - | 2, 3 | 188 |
