siRNA Details
Gene Symbol: Cul7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025611.5 | NP_079887.3 | 5 | 1671 | ||
GenBank mRNA | AK082134.1 | - | 1 | 3467 | ||
AK147156.1 | - | 5 | 1429 | |||
AK147905.1 | - | 5 | 1387 | |||
AK155454.1 | - | 4 | 2158 | |||
AK155583.1 | - | 5 | 1675 | |||
AK166135.1 | - | 4 | 1998 | |||
AK170580.1 | - | 5 | 1674 | |||
AK170808.1 | - | 5 | 1445 | |||
AK172882.1 | - | 4 | 744 | |||
BC059865.1 | - | 2 | 391 | |||
BC065096.1 | - | 4 | 1307 | |||
BC138998.1 | - | 5 | 1609 | 8862020 | ||
BC139000.1 | - | 5 | 1609 | 8862022 |
