siRNA Details
Gene Symbol: Sike1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025679.3 | NP_079955.1 | 2, 3 | 340 | ||
GenBank mRNA | AK014113.1 | - | 2, 3 | 341 | ||
AK014247.1 | - | 2, 3 | 347 | |||
AK017452.1 | - | 2, 3 | 340 | |||
AK017694.1 | - | 2, 3 | 336 | |||
AK019264.1 | - | 2, 3 | 326 | |||
AK033553.1 | - | 2, 3 | 344 | |||
AK035590.1 | - | 2, 3 | 354 | |||
BC004090.1 | - | 2, 3 | 345 | 3593790 |
