siRNA Details
Gene Symbol: Sike1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025679.3 | NP_079955.1 | 5 | 623 | ||
GenBank mRNA | AK014113.1 | - | 5 | 624 | ||
AK014247.1 | - | 5 | 630 | |||
AK017452.1 | - | 5 | 623 | |||
AK017694.1 | - | 5 | 619 | |||
AK019264.1 | - | 5 | 609 | |||
AK033553.1 | - | 5 | 627 | |||
AK035590.1 | - | 5 | 637 | |||
BC004090.1 | - | 5 | 628 | 3593790 |
