siRNA Details
Gene Symbol: Sike1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025679.3 | NP_079955.1 | 4 | 537 | ||
GenBank mRNA | AK014113.1 | - | 4 | 538 | ||
AK014247.1 | - | 4 | 544 | |||
AK017452.1 | - | 4 | 537 | |||
AK017694.1 | - | 4 | 533 | |||
AK019264.1 | - | 4 | 523 | |||
AK033553.1 | - | 4 | 541 | |||
AK035590.1 | - | 4 | 551 | |||
AK038295.1 | - | 2 | 1724 | |||
BC004090.1 | - | 4 | 542 | 3593790 |
