siRNA Details
Gene Symbol: Tmem186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025708.4 | NP_079984.2 | 2 | 484 | ||
GenBank mRNA | AK009236.1 | - | 2 | 321 | ||
AK013147.1 | - | 2 | 314 | |||
AK014478.1 | - | 2 | 485 | |||
AK033686.1 | - | 2 | 322 | |||
AK088656.1 | - | 2 | 494 | |||
AK159727.1 | - | 2 | 470 | |||
AK159782.1 | - | 2 | 470 | |||
BC027261.1 | - | 2 | 314 | 2649343 | ||
BC058268.1 | - | 1 | 1546 |
