siRNA Details
Gene Symbol: Aspn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172481.1 | NP_001165952.1 | 8 | 1270 | ||
NM_025711.3 | NP_079987.2 | 8 | 1287 | |||
GenBank mRNA | AF316825.1 | - | 7 | 1007 | ||
AK014504.1 | - | 8 | 1243 | |||
AK085920.1 | - | 8 | 1243 | |||
BC034888.1 | - | 8 | 1189 | 1365428 | ||
BC145904.1 | - | 8 | 1185 | 40131207 | ||
BC145930.1 | - | 8 | 1168 | 40131233 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001014008, XM_008771557, XM_006253695, XM_006253707, BC083711.
Gene Ontology Categories:
