siRNA Details
Gene Symbol: Lonp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168591.1 | NP_001162063.1 | 9 | 1379 | ||
NM_025827.3 | NP_080103.1 | 15 | 2683 | |||
GenBank mRNA | AK003766.1 | - | 3 | 520 | ||
AK004843.1 | - | 15 | 2597 | |||
AK047786.1 | - | 756 | ||||
AK050231.1 | - | 15 | 2592 | |||
AK075719.1 | - | 9 | 1380 | |||
AK088348.1 | - | 14 | 2589 | |||
AK151103.1 | - | 15 | 2629 | |||
AK151406.1 | - | 13 | 2197 | |||
AK152964.1 | - | 13 | 2199 | |||
AK168628.1 | - | 13 | 2207 | |||
AK168791.1 | - | 15 | 2629 | |||
AK168956.1 | - | 15 | 2629 | |||
AK184483.1 | - | 1 | 102 | |||
AK197630.1 | - | 1 | 102 | |||
BC049090.1 | - | 15 | 2619 | 6401542 |
