siRNA Details
Gene Symbol: Lonp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168591.1 | NP_001162063.1 | 8 | 1070 | ||
NM_025827.3 | NP_080103.1 | 14 | 2374 | |||
GenBank mRNA | AK003766.1 | - | 2 | 211 | ||
AK004843.1 | - | 14 | 2288 | |||
AK050231.1 | - | 14 | 2283 | |||
AK075719.1 | - | 8 | 1071 | |||
AK088348.1 | - | 13 | 2280 | |||
AK151103.1 | - | 14 | 2320 | |||
AK151406.1 | - | 12 | 1888 | |||
AK152964.1 | - | 12 | 1890 | |||
AK168628.1 | - | 12 | 1898 | |||
AK168791.1 | - | 14 | 2320 | |||
AK168956.1 | - | 14 | 2320 | |||
BC049090.1 | - | 14 | 2310 | 6401542 |
