siRNA Details
Gene Symbol: Pccb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025835.2 | NP_080111.1 | 13, 14 | 1432 | ||
GenBank mRNA | AF327060.1 | - | 13, 14 | 1420 | ||
AK004468.1 | - | 1, 2 | 45 | |||
AK004976.1 | - | 13, 14 | 1433 | |||
AK050366.1 | - | 13, 14 | 1438 | |||
AK134437.1 | - | 12, 13 | 1335 | |||
BC026531.1 | - | 13, 14 | 1408 | |||
BC037082.1 | - | 13, 14 | 1434 | 5098790 | ||
BC055946.1 | - | 13, 14 | 1439 | 4166700 |
