siRNA Details
Gene Symbol: Pccb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025835.2 | NP_080111.1 | 11 | 1200 | ||
GenBank mRNA | AF327060.1 | - | 11 | 1188 | ||
AK004976.1 | - | 11 | 1201 | |||
AK050366.1 | - | 11 | 1206 | |||
AK134437.1 | - | 10 | 1103 | |||
BC026531.1 | - | 11 | 1176 | |||
BC037082.1 | - | 11 | 1202 | 5098790 | ||
BC055946.1 | - | 11 | 1207 | 4166700 |

More Information
Other Species Targeted

Gene Ontology Categories:
