siRNA Details
Gene Symbol: Prl3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025896.2 | NP_080172.1 | 5 | 824 | ||
GenBank mRNA | AF525154.1 | - | 5 | 809 | ||
AK005474.1 | - | 5 | 825 | |||
AK005567.1 | - | 5 | 822 | |||
BC100334.1 | - | 5 | 761 | 30791042 |
