siRNA Details
Gene Symbol: D16Ertd472e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252438.1 | NP_001239367.1 | 3 | 725 | ||
NM_001252439.1 | NP_001239368.1 | 3 | 492 | |||
NM_001252440.1 | NP_001239369.1 | 3 | 457 | |||
NM_025967.4 | NP_080243.3 | 3 | 525 | |||
GenBank mRNA | AK005829.1 | - | 3 | 436 | ||
AK009258.1 | - | 3 | 696 | |||
AK044670.1 | - | 3 | 526 | |||
BC019957.1 | - | 3 | 416 | 3966397 | ||
BG078224.2 | - | 3 | 417 | |||
CN662712.1 | - | 3 | 484 |
