siRNA Details
Gene Symbol: Mastl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025979.4 | NP_080255.3 | 8 | 1778 | ||
GenBank mRNA | AK006448.1 | - | 3 | 879 | ||
AK012597.1 | - | 3 | 879 | |||
AK030140.1 | - | 8 | 1673 | |||
BC066834.1 | - | 1035 | ||||
BC086483.1 | - | 1047 | ||||
BC103779.1 | - | 8 | 1654 | 30480071 |
