siRNA Details
Gene Symbol: Smarca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011416.2 | NP_035546.2 | 28 | 4266 | ||
NM_026003.2 | NP_080279.1 | 4 | 415 | |||
GenBank mRNA | AK011935.1 | - | 4 | 360 | ||
AK147322.1 | - | 28 | 4467 | |||
BC004653.1 | - | 1 | 19 | |||
BC022975.1 | - | 6 | 892 | |||
BC056199.1 | - | 18 | 2364 | |||
BC075641.1 | - | 28 | 4248 | 30544092 | ||
BY262171.1 | - | 4 | 417 |
