siRNA Details
Gene Symbol: Smarca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011416.2 | NP_035546.2 | 28 | 4159 | ||
NM_026003.2 | NP_080279.1 | 4 | 308 | |||
GenBank mRNA | AK011935.1 | - | 4 | 253 | ||
AK135871.1 | - | 12 | 1510 | |||
AK147322.1 | - | 28 | 4360 | |||
BC022975.1 | - | 6 | 785 | |||
BC055841.1 | - | 2 | 59 | |||
BC056199.1 | - | 18 | 2257 | |||
BC075641.1 | - | 28 | 4141 | 30544092 | ||
BY262171.1 | - | 4 | 310 |
