siRNA Details
Gene Symbol: Zmynd19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026021.3 | NP_080297.1 | 5 | 633 | ||
GenBank mRNA | AJ409150.1 | - | 5 | 594 | ||
AJ409151.1 | - | 5 | 479 | |||
AK005908.1 | - | 5 | 621 | |||
AK012484.1 | - | 5 | 634 | |||
AK088063.1 | - | 5 | 654 | |||
BC027321.1 | - | 5 | 599 | 3990966 |
