siRNA Details
Gene Symbol: Mboat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083341.1 | NP_001076810.1 | 7 | 833 | ||
NM_026037.3 | NP_080313.2 | 8 | 929 | |||
GenBank mRNA | AB297383.1 | - | 8 | 828 | ||
AB305046.1 | - | 8 | 828 | |||
AK012931.1 | - | 7 | 817 | |||
AK031824.1 | - | 7 | 818 | |||
AK033106.1 | - | 5 | 593 | |||
AK034873.1 | - | 5 | 617 | |||
AK076045.1 | - | 7 | 823 | |||
BC025020.1 | - | 7 | 783 | 5364068 |
