siRNA Details
Gene Symbol: 2810474O19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289661.1 | NP_001276590.1 | 4 | 3076 | ||
NM_001289662.1 | NP_001276591.1 | 3 | 2989 | |||
NM_026054.3 | NP_080330.1 | 3 | 3007 | |||
GenBank mRNA | AK031526.1 | - | 3 | 2870 | ||
AK031989.1 | - | 4 | 3039 | |||
AK079405.1 | - | 4 | 2907 | |||
AK220403.1 | - | 3 | 2990 | |||
BC023168.1 | - | 1 | 983 | |||
BC086679.1 | - | 1 | 2427 | |||
BC139435.1 | - | 3 | 2929 | 8862457 | ||
BC139436.1 | - | 4 | 3016 | 8862458 |
