siRNA Details
Gene Symbol: 5730559C18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028872.3 | NP_083148.3 | 3 | 542 | ||
GenBank mRNA | AK017848.1 | - | 2 | 182 | ||
BB611110.1 | - | 3 | 542 | |||
BC052416.1 | - | 3 | 526 | 6402346 | ||
BC053100.1 | - | 3 | 526 | 6402346 |
