siRNA Details
Gene Symbol: Herc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026101.4 | NP_080377.2 | 15 | 2129 | ||
NM_030114.2 | NP_084390.1 | 15 | 2129 | |||
GenBank mRNA | AK143111.1 | - | 15 | 2003 | ||
AK145735.1 | - | 10 | 1077 | |||
AK147723.1 | - | 15 | 2016 | |||
BC043082.1 | - | 15 | 1941 | 6417111 | ||
BC047157.1 | - | 1 | 31 | |||
BC060033.1 | - | 13 | 1714 | 6478123 |
