siRNA Details
Gene Symbol: Herc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026101.4 | NP_080377.2 | 25 | 3582 | ||
NM_030114.2 | NP_084390.1 | 26 | 3606 | |||
GenBank mRNA | AK019546.1 | - | 23 | 3214 | ||
AK143111.1 | - | 26 | 3480 | |||
AK145735.1 | - | 20 | 2530 | |||
AK147723.1 | - | 25 | 3469 | |||
BC026855.1 | - | 11 | 1339 | |||
BC043082.1 | - | 25 | 3394 | 6417111 | ||
BC047157.1 | - | 11 | 1484 | |||
BC060033.1 | - | 24 | 3191 | 6478123 |
