siRNA Details
Gene Symbol: 1700093K21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110133.1 | NP_001103603.1 | 5 | 943 | ||
NM_026105.3 | NP_080381.2 | 5 | 625 | |||
GenBank mRNA | AK006386.1 | - | 2 | 129 | ||
AK007052.1 | - | 5 | 531 | |||
AK007139.1 | - | 4 | 382 | |||
BC049567.1 | - | 5 | 625 | 6773879 |
