siRNA Details
Gene Symbol: 2210010C04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023333.4 | NP_075822.3 | 1 | 47 | ||
GenBank mRNA | AK008460.1 | - | 1 | 48 | ||
AK008695.1 | - | 1 | 46 | |||
BC061093.1 | - | 1 | 48 | 30306963 |
