siRNA Details
Gene Symbol: Bbs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026116.2 | NP_080392.1 | 7 | 1194 | ||
GenBank mRNA | AF342737.1 | - | 7 | 1194 | ||
AK010779.1 | - | 7 | 807 | |||
BC057184.1 | - | 7 | 853 | 5066846 |
