siRNA Details
Gene Symbol: Pkp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026163.2 | NP_080439.1 | 12 | 2290 | ||
GenBank mRNA | AK004650.1 | - | 12 | 2291 | ||
AK005020.1 | - | 12 | 2322 | |||
AK049481.1 | - | 12 | 2291 | |||
AK167660.1 | - | 12 | 2292 | |||
BC018487.1 | - | 6 | 846 | |||
BC037133.1 | - | 7 | 896 | |||
BC068279.1 | - | 10 | 1571 | |||
BC138252.1 | - | 12 | 2290 | 8861271 | ||
BC138253.1 | - | 12 | 2290 | 8861272 |
