siRNA Details
Gene Symbol: Decr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026172.3 | NP_080448.1 | 8 | 916 | ||
GenBank mRNA | AK002756.1 | - | 8 | 796 | ||
AK004725.2 | - | 8 | 783 | |||
AK015692.1 | - | 8 | 787 | |||
AK146003.1 | - | 8 | 787 | |||
AK168490.1 | - | 8 | 784 | |||
BC046972.1 | - | 8 | 916 | 6469277 | ||
CT010405.1 | - | 8 | 760 |
