siRNA Details
Gene Symbol: 1700019N19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026208.2 | NP_080484.1 | 5 | 532 | ||
GenBank mRNA | AK006135.1 | - | 5 | 533 | ||
AK006423.1 | - | 5 | 529 | |||
BC049561.1 | - | 5 | 520 | 6773246 |
