siRNA Details
Gene Symbol: Lrrc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146021.1 | NP_001139493.1 | 3 | 904 | ||
NM_026253.4 | NP_080529.1 | 3 | 904 | |||
GenBank mRNA | AK005666.1 | - | 3 | 844 | ||
AK006063.1 | - | 3 | 841 | |||
AK014952.1 | - | 3 | 869 | |||
AK015365.1 | - | 3 | 908 | |||
AK029909.1 | - | 3 | 869 | |||
AY775400.1 | - | 3 | 973 | |||
BC048444.1 | - | 3 | 889 | |||
BC061072.1 | - | 1 | 632 | 6771444 |
