siRNA Details
Gene Symbol: Luc7l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026313.1 | NP_080589.1 | 3, 4 | 337 | ||
GenBank mRNA | AK014362.1 | - | 3, 4 | 330 | ||
AK019464.1 | - | 3, 4 | 390 | |||
AK078090.1 | - | 3, 4 | 384 | |||
AK151228.1 | - | 3, 4 | 385 | |||
AK151839.1 | - | 3, 4 | 385 | |||
BC009092.1 | - | 3, 4 | 337 | 3157532 | ||
EF410121.1 | - | 369 |
