siRNA Details
Gene Symbol: Reg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026328.2 | NP_080604.2 | 5, 6 | 571 | ||
GenBank mRNA | AK008049.1 | - | 5, 6 | 573 | ||
AK008438.1 | - | 5, 6 | 574 | |||
AK160329.1 | - | 5, 6 | 569 | |||
AK162316.1 | - | 5, 6 | 572 | |||
BC019465.1 | - | 5, 6 | 554 | 4216795 |
