siRNA Details
Gene Symbol: Snap47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144521.2 | NP_653104.1 | 2 | 112 | ||
GenBank mRNA | AK028053.1 | - | 2 | 128 | ||
AK049330.1 | - | 2 | 156 | |||
AK075640.1 | - | 2 | 151 | |||
AK081883.1 | - | 2 | 128 | |||
AK144165.1 | - | 2 | 165 | |||
BC023182.1 | - | 2 | 112 | 4949809 | ||
BC029025.1 | - | 2 | 122 | 5036036 |
