siRNA Details
Gene Symbol: Uxs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026430.3 | NP_080706.1 | 8, 9 | 613 | ||
GenBank mRNA | AF399958.1 | - | 8, 9 | 613 | ||
AK005536.1 | - | 8, 9 | 553 | |||
AK075806.1 | - | 8, 9 | 665 | |||
AK076166.1 | - | 7, 8 | 654 | |||
AK152376.1 | - | 8, 9 | 668 | |||
AK208821.1 | - | 3, 4 | 353 | |||
BC037049.1 | - | 8, 9 | 659 | 4935237 |
