siRNA Details
Gene Symbol: Zfp169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164575.1 | NP_001158047.1 | 5 | 671 | ||
NM_026450.3 | NP_080726.3 | 5 | 703 | |||
GenBank mRNA | BC060222.1 | - | 5 | 661 | 30356624 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK015230.1 | - | 79 | Chr. - - [-] on Build |
