siRNA Details
Gene Symbol: Gnpda2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038015.1 | NP_001033104.1 | 6 | 771 | ||
GenBank mRNA | AK015527.1 | - | 6 | 781 | ||
AK016785.1 | - | 820 | ||||
AK017588.1 | - | 6 | 780 | |||
AK051596.1 | - | 6 | 780 | |||
AK141408.1 | - | 6 | 783 | |||
AK166601.1 | - | 6 | 773 | |||
AK168516.1 | - | 6 | 790 | |||
AK171299.1 | - | 6 | 766 | |||
AK220275.1 | - | 6 | 3333 | |||
BC004084.1 | - | 6 | 771 | 3593102 |
