siRNA Details
Gene Symbol: Srsf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026499.4 | NP_080775.3 | 5 | 805 | ||
GenBank mRNA | AK004831.1 | - | 5 | 786 | ||
AK012605.1 | - | 5 | 553 | |||
AK051608.1 | - | 3 | 958 | |||
AK133872.1 | - | 5 | 786 | |||
AK146293.1 | - | 5 | 783 | |||
AK159519.1 | - | 5 | 783 | |||
AK171709.1 | - | 4 | 2069 | |||
BC012039.1 | - | 5 | 792 | 2648111 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001014185, BC087121, FM081417.
Gene Ontology Categories:
