siRNA Details
Gene Symbol: 9430016H08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081181.2 | NP_001074650.1 | 2 | 702 | ||
GenBank mRNA | AK007831.1 | - | 2 | 491 | ||
AK079116.1 | - | 2 | 704 | |||
AK165130.1 | - | 1 | 3186 | |||
BC024945.1 | - | 2 | 171 | 3989472 | ||
BC071241.1 | - | 499 | 5125554 |
